Assay Details
| Gene Symbol: Chst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145142.2 | NP_660124.2 | 2 - 3 | 389 | 79 | |
| GenBank mRNA | AF360543.1 | - | 2 - 3 | 221 | 79 | |
| AK046415.1 | - | 2 - 3 | 241 | 79 | ||
| AK047378.1 | - | 2 - 3 | 383 | 79 | ||
| BC056956.1 | - | 2 - 3 | 318 | 79 | ||
| BC132223.1 | - | 1 - 2 | 61 | 40130500 | 79 | |
| BC132225.1 | - | 1 - 2 | 61 | 40130502 | 79 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF360543.1, AK046415.1, AK047378.1, BC132223.1, BC132225.1, NM_145142.2 transcripts.
Set Membership: |
|

Chst10
Genomic Map