Assay Details
| Gene Symbol: Chst10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145142.2 | NP_660124.2 | 4 - 5 | 610 | 71 | |
| GenBank mRNA | AF360543.1 | - | 4 - 5 | 442 | 71 | |
| AK046415.1 | - | 4 - 5 | 462 | 71 | ||
| AK047378.1 | - | 5 - 6 | 734 | 71 | ||
| AK050676.1 | - | 3 - 4 | 492 | 71 | ||
| BC026960.1 | - | 2 - 3 | 119 | 71 | ||
| BC056956.1 | - | 4 - 5 | 539 | 71 | ||
| BC132223.1 | - | 3 - 4 | 282 | 40130500 | 71 | |
| BC132225.1 | - | 3 - 4 | 282 | 40130502 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chst10
Genomic Map