| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700004423.mm.1 |
TC0700000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004423.mm.1 |
NM_001174155.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004423.mm.1 |
NM_001174155.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004423.mm.1 |
NM_145149.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004423.mm.1 |
NM_145149.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004423.mm.1 |
NR_045676.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004423.mm.1 |
NR_045676.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004422.mm.1 |
TC0700000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004422.mm.1 |
NM_001174155.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004422.mm.1 |
NM_001174155.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004422.mm.1 |
NM_145149.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004422.mm.1 |
NM_145149.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004422.mm.1 |
NR_045676.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004422.mm.1 |
NR_045676.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004424.mm.1 |
TC0700000522.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700004424.mm.1 |
NM_001174155.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004424.mm.1 |
NM_001174155.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004424.mm.1 |
NM_145149.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004424.mm.1 |
NM_145149.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004424.mm.1 |
NR_045676.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700004424.mm.1 |
NR_045676.1 |
Refseq |