Assay Details
| Gene Symbol: Abhd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190437.1 | NP_001177366.1 | 4 - 5 | 547 | 72 | |
| NM_145215.2 | NP_660250.1 | 4 - 5 | 1024 | 72 | ||
| NR_033794.1 | 3 - 4 | 1009 | 72 | |||
| GenBank mRNA | AF412033.1 | - | 4 - 5 | 1024 | 72 | |
| AK004244.1 | - | 4 - 5 | 558 | 72 | ||
| BC016241.1 | - | 2 - 3 | 598 | 72 | ||
| BC038052.1 | - | 3 - 4 | 426 | 72 | ||
| BC051486.1 | - | 2 - 3 | 72 | |||
| BC056610.1 | - | 4 - 5 | 686 | 72 | ||
| BC056624.1 | - | 3 - 4 | 1009 | 72 | ||
| BC069866.1 | - | 4 - 5 | 646 | 6478980 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd11
Genomic Map