Assay Details
| Gene Symbol: Gps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177874.1 | NP_001171345.1 | 4 - 5 | 514 | 81 | |
| NM_145370.2 | NP_663345.2 | 4 - 5 | 654 | 81 | ||
| GenBank mRNA | AK088490.1 | - | 4 - 5 | 511 | 81 | |
| BC003350.1 | - | 4 - 5 | 654 | 3481979 | 81 | |
| BC103782.1 | - | 4 - 5 | 481 | 6486599 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gps1
Genomic Map