Assay Details
| Gene Symbol: Ube2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145420.2 | NP_663395.1 | 2 - 3 | 229 | 110 | |
| GenBank mRNA | AK043167.1 | - | 2 - 3 | 185 | 110 | |
| AK144468.1 | - | 2 - 3 | 184 | 110 | ||
| AK148364.1 | - | 2 - 3 | 217 | 110 | ||
| BC019464.1 | - | 2 - 3 | 200 | 4205941 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2d1
Genomic Map