Assay Details
| Gene Symbol: Smox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177833.1 | NP_001171304.1 | 5 - 6 | 1569 | 76 | |
| NM_001177836.1 | NP_001171307.1 | 6 - 7 | 1161 | 76 | ||
| NM_001177837.1 | NP_001171308.1 | 5 - 6 | 1059 | 76 | ||
| NM_145533.2 | NP_663508.1 | 5 - 6 | 1569 | 76 | ||
| GenBank mRNA | AF495852.1 | - | 6 - 7 | 1030 | 76 | |
| AF495853.1 | - | 5 - 6 | 1438 | 76 | ||
| AJ567473.1 | - | 5 - 6 | 1488 | 76 | ||
| AJ567479.1 | - | 5 - 6 | 845 | 76 | ||
| AJ567480.1 | - | 4 - 5 | 1631 | 76 | ||
| AK143129.1 | - | 5 - 6 | 1543 | 76 | ||
| AK159275.1 | - | 5 - 6 | 1496 | 76 | ||
| AK159372.1 | - | 5 - 6 | 1496 | 76 | ||
| AK166350.1 | - | 958 | 76 | |||
| BC004831.1 | - | 5 - 6 | 1488 | 2647695 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smox
Genomic Map