Assay Details
| Gene Symbol: Ahcyl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145542.3 | NP_663517.2 | 2 - 3 | 502 | 70 | |
| GenBank mRNA | AB092504.1 | - | 2 - 3 | 231 | 70 | |
| BC018218.1 | - | 2 - 3 | 331 | 4007102 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ahcyl1
Genomic Map