Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700020972.mm.1 |
TC0700002360.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020972.mm.1 |
NM_001168469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020972.mm.1 |
NM_001168469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020968.mm.1 |
TC0700002360.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020968.mm.1 |
NM_001168469.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020968.mm.1 |
NM_001168469.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020968.mm.1 |
NM_145578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020968.mm.1 |
NM_145578.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020969.mm.1 |
TC0700002360.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020969.mm.1 |
NM_145578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020969.mm.1 |
NM_145578.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020970.mm.1 |
TC0700002360.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020970.mm.1 |
NM_145578.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020970.mm.1 |
NM_145578.3 |
Refseq |