Assay Details
Gene Symbol: Igflr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145580.2 | NP_663555.2 | 2 - 3 | 258 | 67 | |
GenBank mRNA | AK154129.1 | - | 2 - 3 | 259 | 67 | |
BC019733.1 | - | 1 - 2 | 3676256 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955078 | BC019733.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955078 | NM_145580.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955078 | AI661657.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955078 | AK154129.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 923172 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 150051 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136316 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955078 | BC019733.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955078 | NM_145580.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955078 | AI661657.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955078 | AK154129.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032230 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032232 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032218 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032210 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032228 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032229 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032212 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032227 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032222 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032220 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032214 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032231 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032223 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032224 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032226 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25032217 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136316 | - | - |
Whole Mouse Genome G4122A | A_52_P640413 | BC019733.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131220 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136316 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06035633 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06024152 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06024158 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032212 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032169 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032195 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032170 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032188 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25032217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06024143 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032214 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032228 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06024147 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06024158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06024146 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25032218 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10551993 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551995 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551994 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551987 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551990 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551988 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551992 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551991 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10551985 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1426126_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1441880_PM_x_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10551989 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10551981 | - | - |
Mouse Genome 430 2.0 Array | 1434305_at | - | - |
Mouse Genome 430 2.0 Array | 1441880_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1434305_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1426126_a_at | - | - |
Mouse Genome 430 2.0 Array | 1441880_x_at | - | - |
Mouse Genome 430 2.0 Array | 1426126_PM_a_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101883_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101883_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700005072.mm.1 | TC0700000567.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700005074.mm.1 | TC0700000567.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700005074.mm.1 | NM_145580.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700005074.mm.1 | NM_145580.2 | Refseq |
Mouse Expression Set 430 | 1434305_at | - | - |
Mouse Expression Set 430 | 1426126_a_at | - | - |
Mouse Expression Set 430 | 1426126_PM_a_at | - | - |
Mouse Expression Set 430 | 1434305_PM_at | - | - |
Mouse Expression Set 430 | 1441880_x_at | - | - |
Mouse Expression Set 430 | 1441880_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1434305_at | - | - |
Mouse Genome 430A 2.0 Array | 1426126_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1426126_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1434305_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426126_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1426126_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441880_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1441880_x_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 681688 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6420068 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006420068 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244377 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244377 | - | - |
mouse-6 v1.1 (Illumina) | scl0101883.3_27-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6420068 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0101883.3_27-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420068 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1244377 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244377 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0101883.3_27-S | - | - |
