Assay Details
| Gene Symbol: Dkk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145592.2 | NP_663567.1 | 2 - 3 | 404 | 59 | |
| GenBank mRNA | BC018400.1 | - | 2 - 3 | 404 | 3675956 | 59 |
More Information
Updated Mapping Information
Set Membership: |
|

Dkk4
Genomic Map