Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00013790.mm.1 |
NM_001271447.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013790.mm.1 |
NM_001271448.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013790.mm.1 |
NM_001271449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013790.mm.1 |
NM_001271450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013790.mm.1 |
NM_001271451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013790.mm.1 |
NM_145951.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013790.mm.1 |
TC0X00002171.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00013791.mm.1 |
NM_001271447.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013791.mm.1 |
NM_001271448.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013791.mm.1 |
NM_001271449.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013791.mm.1 |
NM_001271450.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013791.mm.1 |
NM_001271451.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013791.mm.1 |
NM_145951.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00013791.mm.1 |
TC0X00002171.mm.1 |
MTA TC |