Assay Details
| Gene Symbol: Cth | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145953.2 | NP_666065.1 | 10 - 11 | 1081 | 60 | |
| GenBank mRNA | AK002480.1 | - | 10 - 11 | 1059 | 60 | |
| AY083352.1 | - | 10 - 11 | 1081 | 60 | ||
| BC019483.1 | - | 10 - 11 | 1067 | 4235178 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cth
Genomic Map