Assay Details
| Gene Symbol: Phyhip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145981.3 | NP_666093.1 | 1 - 2 | 285 | 88 | |
| GenBank mRNA | AK049900.1 | - | 1 - 2 | 286 | 88 | |
| BC030494.1 | - | 1 - 2 | 282 | 88 | ||
| BC139360.1 | - | 1 - 3 | 8862382 | 88 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK049900.1,BC139360.1,NM_145981.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368210 | BC030494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368210 | NM_145981.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368210 | AK049900.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368210 | BC030494.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368210 | NM_145981.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P368210 | AK049900.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821362 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450894 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571082 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 58091 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 428143 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435987 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 81736 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P150378 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368210 | BC030494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368210 | NM_145981.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368210 | AK049900.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368210 | BC030494.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368210 | NM_145981.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P368210 | AK049900.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368210 | BC030494.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368210 | NM_145981.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368210 | AK049900.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368210 | BC030494.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368210 | NM_145981.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P368210 | AK049900.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P130165 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120529 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11401079 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11401078 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11401066 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11401067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701319 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701373 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P29701342 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11554110 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701362 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11401056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701455 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11128458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701334 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11644966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701339 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11374448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701336 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701374 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11401070 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701407 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701443 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11235092 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11825255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11752842 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11521411 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701386 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701360 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701403 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11825260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701352 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701345 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701373 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701357 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701324 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701368 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701327 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701331 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11252558 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P29701340 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11711742 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10416276 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416277 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416278 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416274 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10416275 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455463_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10416273 | - | - |
| Mouse Genome 430 2.0 Array | 1455463_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455463_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105653_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105653_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1400008001.mm.1 | NM_145981.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008001.mm.1 | NM_145981.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008001.mm.1 | TC1400000998.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1400008003.mm.1 | NM_145981.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008003.mm.1 | NM_145981.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1400008003.mm.1 | TC1400000998.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1455463_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1455463_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 559730 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1170377 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1170377 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001170377 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223321 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223321 | - | - |
| mouse-6 v1.1 (Illumina) | scl46125.6_409-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1170377 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46125.6_409-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1170377 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223321 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223321 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl46125.6_409-S | - | - |

Phyhip
Genomic Map