Assay Details
| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146073.3 | NP_666185.3 | 4 - 5 | 1546 | 145 | |
| GenBank mRNA | AF542387.1 | - | 5 - 6 | 1215 | 145 | |
| AK041496.1 | - | 4 - 5 | 1229 | 145 | ||
| AK041648.1 | - | 5 - 6 | 1542 | 145 | ||
| AK046719.1 | - | 4 - 5 | 1534 | 145 | ||
| AK080845.1 | - | 4 - 5 | 1550 | 145 | ||
| BC059814.1 | - | 4 - 5 | 1589 | 6813525 | 145 | |
| BC086758.1 | - | 5 - 6 | 602 | 145 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc14
Genomic Map