Assay Details
| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146073.3 | NP_666185.3 | 5 - 6 | 1601 | 69 | |
| GenBank mRNA | AF542387.1 | - | 6 - 7 | 1270 | 69 | |
| AK041496.1 | - | 5 - 6 | 1284 | 69 | ||
| AK041648.1 | - | 6 - 7 | 1597 | 69 | ||
| AK046719.1 | - | 5 - 6 | 1589 | 69 | ||
| AK080845.1 | - | 5 - 6 | 1605 | 69 | ||
| BC059814.1 | - | 5 - 6 | 1644 | 6813525 | 69 | |
| BC086758.1 | - | 6 - 7 | 657 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc14
Genomic Map