Assay Details
| Gene Symbol: Zdhhc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146073.3 | NP_666185.3 | 7 - 8 | 1818 | 74 | |
| GenBank mRNA | AF542387.1 | - | 8 - 9 | 1487 | 74 | |
| AK041496.1 | - | 7 - 8 | 1501 | 74 | ||
| AK046719.1 | - | 7 - 8 | 1806 | 74 | ||
| AK080845.1 | - | 7 - 8 | 1822 | 74 | ||
| BC059814.1 | - | 7 - 8 | 1861 | 6813525 | 74 | |
| BC086758.1 | - | 8 - 9 | 874 | 74 | ||
More Information
Set Membership: |
|

Zdhhc14
Genomic Map