Assay Details
| Gene Symbol: Slc5a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146198.2 | NP_666310.1 | 12 - 13 | 1385 | 64 | |
| GenBank mRNA | AF518726.1 | - | 13 - 14 | 1478 | 64 | |
| AK008219.1 | - | 12 - 13 | 1440 | 64 | ||
| BC031742.1 | - | 12 - 13 | 1386 | 4221537 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a11
Genomic Map