Assay Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146207.2 | NP_666319.2 | 1 - 2 | 182 | 78 | |
| GenBank mRNA | AK144802.1 | - | 1 - 2 | 164 | 78 | |
| AK150237.1 | - | 1 - 2 | 161 | 78 | ||
| AK166251.1 | - | 1 - 2 | 164 | 78 | ||
| AK168215.1 | - | 1 - 2 | 164 | 78 | ||
| AK170722.1 | - | 1 - 2 | 164 | 78 | ||
| BC010211.1 | - | 1 - 2 | 160 | 3969515 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cul4a
Genomic Map