Assay Details
| Gene Symbol: Cul4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146207.2 | NP_666319.2 | 4 - 5 | 467 | 76 | |
| GenBank mRNA | AK144802.1 | - | 4 - 5 | 449 | 76 | |
| AK150237.1 | - | 4 - 5 | 446 | 76 | ||
| AK166251.1 | - | 4 - 5 | 449 | 76 | ||
| AK168215.1 | - | 4 - 5 | 449 | 76 | ||
| AK170722.1 | - | 4 - 5 | 449 | 76 | ||
| AK182330.1 | - | 3 - 4 | 227 | 76 | ||
| BC010211.1 | - | 4 - 5 | 445 | 3969515 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cul4a
Genomic Map