Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 9 - 10 | 1349 | 64 | |
| GenBank mRNA | AK031952.1 | - | 9 - 10 | 1384 | 64 | |
| AK043319.1 | - | 9 - 10 | 1381 | 64 | ||
| AK046826.1 | - | 9 - 10 | 1321 | 64 | ||
| AK084520.1 | - | 9 - 10 | 1400 | 64 | ||
| AK140191.1 | - | 9 - 10 | 1376 | 64 | ||
| AK171577.1 | - | 9 - 10 | 1389 | 64 | ||
| AK171883.1 | - | 9 - 10 | 1377 | 64 | ||
| AK185154.1 | - | 1 - 2 | 161 | 64 | ||
| AK187571.1 | - | 1 - 2 | 161 | 64 | ||
| AK190879.1 | - | 1 - 2 | 183 | 64 | ||
| AK197455.1 | - | 1 - 2 | 161 | 64 | ||
| AK198087.1 | - | 1 - 2 | 161 | 64 | ||
| AK199564.1 | - | 1 - 2 | 161 | 64 | ||
| AK199815.1 | - | 1 - 2 | 161 | 64 | ||
| AK200069.1 | - | 1 - 2 | 161 | 64 | ||
| AK202260.1 | - | 1 - 2 | 161 | 64 | ||
| AK203281.1 | - | 1 - 2 | 164 | 64 | ||
| AK203450.1 | - | 1 - 2 | 161 | 64 | ||
| AK206758.1 | - | 1 - 2 | 177 | 64 | ||
| AK209742.1 | - | 1 - 2 | 161 | 64 | ||
| AK211234.1 | - | 1 - 2 | 161 | 64 | ||
| BC032215.1 | - | 2 - 3 | 262 | 64 | ||
| BC052199.1 | - | 9 - 10 | 1349 | 6512678 | 64 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC032215.1 transcripts.
Set Membership: |
|

Vac14
Genomic Map