Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 11 - 12 | 1558 | 67 | |
| GenBank mRNA | AK031952.1 | - | 11 - 12 | 1593 | 67 | |
| AK043319.1 | - | 11 - 12 | 1590 | 67 | ||
| AK046826.1 | - | 11 - 12 | 1530 | 67 | ||
| AK054468.1 | - | 2 - 3 | 385 | 67 | ||
| AK084520.1 | - | 11 - 12 | 1609 | 67 | ||
| AK140191.1 | - | 11 - 12 | 1585 | 67 | ||
| AK171577.1 | - | 11 - 12 | 1598 | 67 | ||
| AK171883.1 | - | 11 - 12 | 1586 | 67 | ||
| AK183924.1 | - | 2 - 3 | 218 | 67 | ||
| BC032215.1 | - | 4 - 5 | 471 | 67 | ||
| BC052199.1 | - | 11 - 12 | 1558 | 6512678 | 67 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC032215.1 transcripts.
Set Membership: |
|

Vac14
Genomic Map