Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 16 - 17 | 2209 | 60 | |
| GenBank mRNA | AK031952.1 | - | 16 - 17 | 2244 | 60 | |
| AK046826.1 | - | 16 - 17 | 2181 | 60 | ||
| AK054468.1 | - | 6 - 7 | 2136 | 60 | ||
| AK084520.1 | - | 16 - 17 | 2324 | 60 | ||
| AK140191.1 | - | 15 - 16 | 2079 | 60 | ||
| AK171577.1 | - | 16 - 17 | 2249 | 60 | ||
| AK171883.1 | - | 16 - 17 | 2237 | 60 | ||
| AK199437.1 | - | 1 - 2 | 31 | 60 | ||
| AK205416.1 | - | 2 - 3 | 288 | 60 | ||
| AK217414.1 | - | 1 - 2 | 31 | 60 | ||
| BC032215.1 | - | 9 - 10 | 1122 | 60 | ||
| BC052199.1 | - | 16 - 17 | 2209 | 6512678 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map