Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 17 - 18 | 2293 | 76 | |
| GenBank mRNA | AK046826.1 | - | 17 - 18 | 2265 | 76 | |
| AK084520.1 | - | 17 - 18 | 2408 | 76 | ||
| AK171577.1 | - | 17 - 18 | 2333 | 76 | ||
| AK171883.1 | - | 17 - 18 | 2321 | 76 | ||
| AK217414.1 | - | 2 - 3 | 115 | 76 | ||
| BC032215.1 | - | 10 - 11 | 1206 | 76 | ||
| BC052199.1 | - | 17 - 18 | 2293 | 6512678 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map