Assay Details
| Gene Symbol: Mettl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146247.1 | NP_666359.1 | 3 - 4 | 555 | 93 | |
| GenBank mRNA | AK089212.1 | - | 3 - 4 | 570 | 93 | |
| AK136614.1 | - | 3 - 4 | 581 | 93 | ||
| BC024814.1 | - | 3 - 4 | 555 | 5359405 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mettl22
Genomic Map