Assay Details
| Gene Symbol: Gpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146250.2 | NP_666362.1 | 2 - 3 | 255 | 117 | |
| GenBank mRNA | BC032934.1 | - | 2 - 3 | 183 | 1363204 | 117 |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC032934.1,NM_146250.2 transcripts.
Set Membership: |
|

Gpr1
Genomic Map