Assay Details
| Gene Symbol: Hpse | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152803.4 | NP_690016.1 | 4 - 5 | 744 | 75 | |
| GenBank mRNA | AF359507.1 | - | 4 - 5 | 733 | 75 | |
| AK040471.1 | - | 4 - 5 | 745 | 75 | ||
| AK087283.1 | - | 4 - 5 | 771 | 75 | ||
| AK154628.1 | - | 4 - 5 | 745 | 75 | ||
| AY077467.1 | - | 4 - 5 | 736 | 75 | ||
| BC138782.1 | - | 4 - 5 | 738 | 8861804 | 75 | |
| BC138784.1 | - | 4 - 5 | 738 | 8861806 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hpse
Genomic Map