Assay Details
| Gene Symbol: Ebf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110513.1 | NP_001103983.1 | 7 - 8 | 711 | 82 | |
| GenBank mRNA | AF387630.1 | - | 7 - 8 | 711 | 82 | |
| AF387631.1 | - | 7 - 8 | 711 | 82 | ||
| AF387632.1 | - | 7 - 8 | 711 | 82 | ||
| AF387633.1 | - | 7 - 8 | 711 | 82 | ||
| AF387634.1 | - | 7 - 8 | 711 | 82 | ||
| BC138119.1 | - | 7 - 8 | 711 | 8861138 | 82 | |
| BC145155.1 | - | 8 - 9 | 797 | 9053706 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ebf4
Genomic Map