Assay Details
| Gene Symbol: Mkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082536.1 | NP_001076005.1 | 11 - 12 | 2634 | 118 | |
| NM_153049.3 | NP_694629.2 | 10 - 11 | 2396 | 118 | ||
| GenBank mRNA | AF385582.1 | - | 10 - 11 | 2167 | 118 | |
| AF532597.1 | - | 11 - 12 | 2518 | 118 | ||
| AK044188.1 | - | 11 - 12 | 2627 | 118 | ||
| AK155821.1 | - | 11 - 12 | 2640 | 118 | ||
| AK155937.1 | - | 10 - 11 | 2380 | 118 | ||
| BC050941.1 | - | 11 - 12 | 2527 | 6821370 | 118 | |
| BC054801.1 | - | 11 - 12 | 2570 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mkl1
Genomic Map