Assay Details
Gene Symbol: Zfp472 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153063.3 | NP_694703.2 | 3 - 4 | 283 | 126 | |
GenBank mRNA | AK142395.1 | - | 3 - 4 | 274 | 126 | |
AY195875.1 | - | 3 - 4 | 218 | 126 | ||
BB652931.1 | - | 3 - 4 | 284 | 126 | ||
BC023090.1 | - | 3 - 4 | 218 | 126 | ||
BC027407.1 | - | 3 - 4 | 273 | 4945483 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147863 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147861 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147864 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147862 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | BC027407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | AY195875.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | BC023090.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | NM_153063.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | AK142395.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | BC027407.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | AY195875.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | BC023090.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | NM_153063.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P664617 | AK142395.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137005 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 383932 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501832 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 249460 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 541873 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223700 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919304 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128286 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142643 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | BC027407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | AY195875.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | BC023090.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | NM_153063.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | AK142395.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | BC027407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | AY195875.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | BC023090.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | NM_153063.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P664617 | AK142395.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147840 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147845 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147866 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147841 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147849 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147864 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31147855 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142643 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128286 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08762334 | - | - |
Whole Mouse Genome G4122A | A_51_P470966 | BC027407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664619 | BC027407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664619 | AY195875.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664619 | BC023090.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664619 | NM_153063.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664619 | AB010338.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | BC027407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | AY195875.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | BC023090.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | NM_153063.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | AK142395.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | BC027407.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | AY195875.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | BC023090.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | NM_153063.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P664617 | AK142395.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103414 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128091 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142643 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128286 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13461734 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13250930 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13250937 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13250939 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13250934 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13250941 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147862 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147864 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147861 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31147863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147934 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147864 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147943 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147941 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147828 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147908 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147874 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147903 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147930 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147891 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147830 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147919 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31147884 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13250938 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147869 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13055576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147875 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13250930 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147834 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13544470 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147830 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147859 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13322516 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147849 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13219551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13434483 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13117937 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13250936 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147833 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13389710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31147882 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10443914 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443917 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443915 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443913 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10443916 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425058_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10443912 | - | - |
Mouse Genome 430 2.0 Array | 1425058_at | - | - |
Mouse Genome 430 2.0 Array | 1425058_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224691_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 224691_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1700005198.mm.1 | NM_153063.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005198.mm.1 | NM_153063.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005198.mm.1 | TC1700000571.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700005199.mm.1 | NM_153063.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005199.mm.1 | NM_153063.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700005199.mm.1 | TC1700000571.mm.1 | MTA TC |
Mouse Expression Set 430 | 1425058_at | - | - |
Mouse Expression Set 430 | 1425058_PM_at | - | - |
Murine Genome U74v2 Set | 95285_at | - | - |
Murine Genome U74v2 Set | 95285_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425058_at | - | - |
Mouse Genome 430A 2.0 Array | 1425058_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425058_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425058_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 539629 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1244492 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244492 | - | - |
mouse-6 v1.1 (Illumina) | scl50858.5_334-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1780156 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl50858.5_334-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1780156 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2932722 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1244492 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2932722 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl50858.5_334-S | - | - |
