Assay Details
| Gene Symbol: St7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253703.1 | NP_001240632.1 | 9 - 10 | 1604 | 105 | |
| NM_153091.3 | NP_694731.1 | 11 - 12 | 1838 | 105 | ||
| NR_045578.1 | 11 - 12 | 2104 | 105 | |||
| GenBank mRNA | AB084518.1 | - | 11 - 12 | 1252 | 105 | |
| AK033865.1 | - | 11 - 12 | 1269 | 105 | ||
| AK144075.1 | - | 9 - 10 | 1044 | 105 | ||
| BC024473.1 | - | 11 - 12 | 1229 | 105 | ||
| BC058781.1 | - | 11 - 12 | 1263 | 6311899 | 105 | |
| BC107394.2 | - | 11 - 12 | 1263 | 40058131 | 105 | |
| BC145950.1 | - | 11 - 12 | 1330 | 40131253 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

St7l
Genomic Map