Assay Details
| Gene Symbol: Nupl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153092.4 | NP_694732.3 | 5 - 6 | 726 | 225 | |
| GenBank mRNA | AB067574.1 | - | 4 - 5 | 574 | 148 | |
| AK078478.1 | - | 5 - 6 | 727 | 225 | ||
| BC033270.1 | - | 5 - 6 | 668 | 3981211 | 225 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nupl2
Genomic Map