Assay Details
| Gene Symbol: Cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153107.2 | NP_694747.2 | 3 - 4 | 598 | 65 | |
| GenBank mRNA | AF356844.1 | - | 3 - 4 | 507 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cpz
Genomic Map