Assay Details
| Gene Symbol: Cadm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153112.3 | NP_694752.1 | 4 - 5 | 578 | 63 | |
| GenBank mRNA | AK143204.1 | - | 4 - 5 | 579 | 63 | |
| AY059394.1 | - | 4 - 5 | 502 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cadm4
Genomic Map