Assay Details
| Gene Symbol: Fnbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114665.2 | NP_001108137.1 | 7 - 8 | 799 | 129 | |
| NM_153118.3 | NP_694758.2 | 7 - 8 | 799 | 129 | ||
| GenBank mRNA | BC034510.1 | - | 2 - 3 | 247 | 4481144 | 129 |
More Information
Updated Mapping Information
Set Membership: |
|

Fnbp1l
Genomic Map