| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1300004184.mm.1 |
NM_153131.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004184.mm.1 |
NM_153131.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004184.mm.1 |
TC1300000643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004183.mm.1 |
NM_153131.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004183.mm.1 |
NM_153131.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004183.mm.1 |
TC1300000643.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1300004185.mm.1 |
NM_153131.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004185.mm.1 |
NM_153131.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1300004185.mm.1 |
TC1300000643.mm.1 |
MTA TC |