Assay Details
| Gene Symbol: Unc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153131.3 | NP_694771.1 | 8 - 9 | 1478 | 62 | |
| GenBank mRNA | AJ487852.1 | - | 8 - 9 | 1479 | 62 | |
| AK122575.1 | - | 7 - 8 | 2502 | 62 | ||
| BC058084.1 | - | 7 - 8 | 1331 | 6813463 | 62 | |
More Information
Set Membership: |
|

Unc5a
Genomic Map