Assay Details
| Gene Symbol: Traf3ip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153137.4 | NP_694777.3 | 14 - 15 | 1731 | 104 | |
| GenBank mRNA | AK031400.1 | - | 14 - 15 | 1671 | 104 | |
| AY383616.1 | - | 12 - 13 | 1315 | 104 | ||
| BC036561.1 | - | 14 - 15 | 1539 | 104 | ||
| BC137605.1 | - | 12 - 13 | 1342 | 8860624 | 104 | |
| BC171933.1 | - | 12 - 13 | 1342 | 9054217 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Traf3ip3
Genomic Map