Assay Details
| Gene Symbol: Rab11fip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162868.1 | NP_001156340.1 | 9 - 10 | 2514 | 66 | |
| NM_001162869.1 | NP_001156341.1 | 10 - 11 | 2649 | 66 | ||
| NM_153140.2 | NP_694780.1 | 7 - 8 | 905 | 66 | ||
| GenBank mRNA | AB093257.1 | - | 10 - 11 | 2090 | 66 | |
| AK083658.1 | - | 9 - 10 | 2198 | 66 | ||
| AK085516.1 | - | 7 - 8 | 906 | 66 | ||
| AK128985.1 | - | 7 - 8 | 905 | 66 | ||
| BC023364.1 | - | 7 - 8 | 896 | 66 | ||
| BC037132.1 | - | 7 - 8 | 945 | 66 | ||
More Information
Set Membership: |
|

Rab11fip3
Genomic Map