Assay Details
| Gene Symbol: Rab11fip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162868.1 | NP_001156340.1 | 12 - 13 | 2888 | 134 | |
| NM_001162869.1 | NP_001156341.1 | 13 - 14 | 3023 | 134 | ||
| NM_153140.2 | NP_694780.1 | 10 - 11 | 1279 | 134 | ||
| GenBank mRNA | AB093257.1 | - | 13 - 14 | 2464 | 134 | |
| AK083658.1 | - | 12 - 13 | 2573 | 134 | ||
| BC023364.1 | - | 10 - 11 | 1270 | 134 | ||
| BC037132.1 | - | 10 - 11 | 1319 | 134 | ||
| CB840427.1 | - | 1 - 2 | 174 | 134 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab11fip3
Genomic Map