Assay Details
| Gene Symbol: Txnrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178058.1 | NP_001171529.1 | 3 - 4 | 586 | 61 | |
| NM_001178059.1 | NP_001171530.1 | 3 - 4 | 586 | 61 | ||
| NM_001178060.1 | NP_001171531.1 | 3 - 4 | 586 | 61 | ||
| NM_153162.3 | NP_694802.2 | 3 - 4 | 586 | 61 | ||
| GenBank mRNA | AF349659.1 | - | 3 - 4 | 495 | 61 | |
| AK012699.1 | - | 3 - 4 | 579 | 61 | ||
| AK041046.1 | - | 3 - 4 | 587 | 61 | ||
| AK080362.1 | - | 3 - 4 | 577 | 61 | ||
| AK134987.1 | - | 3 - 4 | 577 | 61 | ||
| BC076605.1 | - | 3 - 4 | 544 | 30628949 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd3
Genomic Map