Assay Details
| Gene Symbol: Txnrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178058.1 | NP_001171529.1 | 10 - 11 | 1455 | 66 | |
| NM_001178059.1 | NP_001171530.1 | 8 - 9 | 1113 | 66 | ||
| NM_001178060.1 | NP_001171531.1 | 8 - 9 | 1113 | 66 | ||
| NM_153162.3 | NP_694802.2 | 10 - 11 | 1455 | 66 | ||
| GenBank mRNA | AF349659.1 | - | 10 - 11 | 1364 | 66 | |
| AK012699.1 | - | 10 - 11 | 1448 | 66 | ||
| AK041046.1 | - | 10 - 11 | 1458 | 66 | ||
| AK080362.1 | - | 10 - 11 | 1446 | 66 | ||
| AK134987.1 | - | 8 - 9 | 1104 | 66 | ||
| BC076605.1 | - | 10 - 11 | 1413 | 30628949 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd3
Genomic Map