Assay Details
| Gene Symbol: Txnrd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001178058.1 | NP_001171529.1 | 14 - 15 | 1905 | 89 | |
| NM_001178059.1 | NP_001171530.1 | 12 - 13 | 1563 | 89 | ||
| NM_001178060.1 | NP_001171531.1 | 12 - 13 | 1563 | 89 | ||
| NM_153162.3 | NP_694802.2 | 14 - 15 | 1905 | 89 | ||
| GenBank mRNA | AF349659.1 | - | 14 - 15 | 1814 | 89 | |
| AK012699.1 | - | 14 - 15 | 1898 | 89 | ||
| AK041046.1 | - | 14 - 15 | 1908 | 89 | ||
| AK080362.1 | - | 14 - 15 | 1896 | 89 | ||
| AK134987.1 | - | 12 - 13 | 1554 | 89 | ||
| AK218006.1 | - | 3 - 4 | 327 | 89 | ||
| BC076605.1 | - | 14 - 15 | 1863 | 30628949 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txnrd3
Genomic Map