Assay Details
| Gene Symbol: Dcaf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153167.2 | NP_694807.2 | 5 - 6 | 1251 | 135 | |
| GenBank mRNA | AK045241.1 | - | 6 - 7 | 1226 | 135 | |
| AK083428.1 | - | 5 - 6 | 852 | 135 | ||
| AK157345.1 | - | 5 - 6 | 1187 | 135 | ||
| AK170534.1 | - | 6 - 7 | 1311 | 135 | ||
| AK194080.1 | - | 2 - 3 | 269 | 135 | ||
| AK213618.1 | - | 2 - 3 | 269 | 135 | ||
| BC027317.1 | - | 5 - 6 | 1211 | 3601889 | 135 | |
| BC085101.1 | - | 5 - 6 | 1108 | 6391172 | 135 | |
| CN533345.1 | - | 3 - 4 | 335 | 135 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf10
Genomic Map