Assay Details
| Gene Symbol: Slc36a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153170.3 | NP_694810.2 | 2 - 3 | 360 | 58 | |
| GenBank mRNA | AF453744.1 | - | 2 - 3 | 313 | 58 | |
| AF512429.1 | - | 2 - 3 | 383 | 58 | ||
| AK041041.1 | - | 2 - 3 | 352 | 58 | ||
| AK079053.1 | - | 2 - 3 | 325 | 58 | ||
| AK086373.1 | - | 2 - 3 | 361 | 58 | ||
| AK156409.1 | - | 2 - 3 | 361 | 58 | ||
| AK160394.1 | - | 2 - 3 | 325 | 58 | ||
| AY228366.1 | - | 2 - 3 | 310 | 58 | ||
| BC044800.1 | - | 2 - 3 | 360 | 4189221 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc36a2
Genomic Map