Assay Details
| Gene Symbol: Ago4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153177.3 | NP_694817.2 | 9 - 10 | 1370 | 72 | |
| GenBank mRNA | AB081474.1 | - | 9 - 10 | 1115 | 72 | |
| AK030018.1 | - | 9 - 10 | 1386 | 72 | ||
| AK129395.2 | - | 6 - 7 | 686 | 72 | ||
| BC096023.1 | - | 9 - 10 | 1152 | 30607807 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ago4
Genomic Map