Assay Details
| Gene Symbol: Pxt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153390.1 | NP_700439.1 | 1 - 2 | 64 | 65 | |
| GenBank mRNA | AF486826.1 | - | 1 - 2 | 64 | 65 | |
| AK005593.1 | - | 1 - 2 | 67 | 65 | ||
| BC100437.1 | - | 1 - 2 | 63 | 30915792 | 65 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF486826.1,BC100437.1,NM_153390.1 transcripts.
Set Membership: |
|

Pxt1
Genomic Map