Assay Details
| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153394.2 | NP_700443.2 | 4 - 5 | 840 | 100 | |
| GenBank mRNA | AF488740.1 | - | 3 - 4 | 364 | 100 | |
| AK141990.1 | - | 4 - 5 | 885 | 100 | ||
| BC059227.1 | - | 4 - 5 | 840 | 6400663 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr156
Genomic Map