Assay Details
| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153394.2 | NP_700443.2 | 6 - 7 | 1079 | 64 | |
| GenBank mRNA | AF488740.1 | - | 5 - 6 | 603 | 64 | |
| AK141990.1 | - | 6 - 7 | 1124 | 64 | ||
| BC059227.1 | - | 6 - 7 | 1079 | 6400663 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr156
Genomic Map