Assay Details
| Gene Symbol: Gpr156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153394.2 | NP_700443.2 | 7 - 8 | 1175 | 62 | |
| GenBank mRNA | AF488740.1 | - | 6 - 7 | 699 | 62 | |
| AK141990.1 | - | 7 - 8 | 1220 | 62 | ||
| BC059227.1 | - | 7 - 8 | 1175 | 6400663 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr156
Genomic Map